Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203716.1 | NP_989047.1 | 8 - 9 | 935 | 78 | |
XM_012962836.2 | XP_012818290.1 | 8 - 9 | 1070 | 78 | ||
GenBank mRNA | BC061354.1 | - | 8 - 9 | 935 | 78 | |
CR761467.2 | - | 8 - 9 | 802 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |