Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203720.1 | NP_989051.1 | 1 - 2 | 174 | 72 | |
GenBank mRNA | BC061341.1 | - | 1 - 2 | 174 | 72 | |
CR762313.2 | - | 1 - 2 | 66 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |