Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203720.1 | NP_989051.1 | 3 - 4 | 371 | 94 | |
GenBank mRNA | BC061341.1 | - | 3 - 4 | 371 | 94 | |
CR762313.2 | - | 3 - 4 | 263 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |