Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tardbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244758.1 | NP_001231687.1 | 3 - 4 | 601 | 104 | |
NM_203723.2 | NP_989054.2 | 3 - 4 | 601 | 104 | ||
XM_012966272.2 | XP_012821726.1 | 3 - 4 | 587 | 104 | ||
XM_012966273.2 | XP_012821727.1 | 3 - 4 | 590 | 104 | ||
GenBank mRNA | BC061336.1 | - | 3 - 4 | 556 | 104 | |
CN089905.1 | - | 3 - 4 | 603 | 104 | ||
CR762161.2 | - | 3 - 4 | 500 | 104 | ||
DN077197.1 | - | 3 - 4 | 500 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |