Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203730.2 | NP_989061.2 | 5 - 6 | 919 | 70 | |
GenBank mRNA | BC061314.1 | - | 5 - 6 | 911 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |