Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: act3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC061264.1 | - | 4 - 5 | 667 | 73 | |
CR760277.2 | - | 3 - 4 | 645 | 73 | ||
CT025518.2 | - | 1466 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |