Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iscu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203757.1 | NP_989088.1 | 1 - 2 | 120 | 81 | |
GenBank mRNA | BC062501.1 | - | 1 - 2 | 120 | 81 | |
CR855562.1 | - | 1 - 2 | 96 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |