Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203777.1 | NP_989108.1 | 5 - 6 | 384 | 98 | |
GenBank mRNA | BC062518.1 | - | 5 - 6 | 384 | 98 | |
CR761977.1 | - | 5 - 6 | 378 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |