Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203801.1 | NP_989132.1 | 4 - 5 | 601 | 103 | |
GenBank mRNA | BC061333.1 | - | 4 - 5 | 601 | 103 | |
CR761233.2 | - | 4 - 5 | 600 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |