Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203823.1 | NP_989154.1 | 66 | 68 | ||
GenBank mRNA | BC061629.1 | - | 66 | 68 | ||
BC080971.1 | - | 66 | 68 | |||
CR942746.2 | - | 61 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |