Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203823.1 | NP_989154.1 | 4 - 5 | 600 | 78 | |
GenBank mRNA | BC061629.1 | - | 4 - 5 | 600 | 78 | |
BC080971.1 | - | 4 - 5 | 600 | 78 | ||
CR942746.2 | - | 4 - 5 | 595 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |