Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vgll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203847.1 | NP_989178.1 | 2 - 3 | 313 | 68 | |
XM_012962842.2 | XP_012818296.1 | 1 - 2 | 458 | 68 | ||
XM_012962843.2 | XP_012818297.1 | 1 - 2 | 463 | 68 | ||
GenBank mRNA | BC063357.1 | - | 2 - 3 | 313 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |