Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203854.1 | NP_989185.1 | 2 - 3 | 171 | 91 | |
GenBank mRNA | BC063201.1 | - | 2 - 3 | 171 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |