Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203919.1 | NP_989250.1 | 3 - 4 | 503 | 80 | |
XM_012964457.2 | XP_012819911.2 | 2 - 3 | 191 | 80 | ||
GenBank mRNA | BC063931.1 | - | 3 - 4 | 503 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |