Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203932.1 | NP_989263.1 | 8 - 9 | 1050 | 62 | |
XM_012969824.2 | XP_012825278.1 | 8 - 9 | 1347 | 62 | ||
GenBank mRNA | BC064248.1 | - | 8 - 9 | 1050 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |