Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203933.1 | NP_989264.1 | 7 - 8 | 1136 | 77 | |
GenBank mRNA | BC064247.1 | - | 7 - 8 | 1136 | 77 | |
BC080335.1 | - | 7 - 8 | 1149 | 77 | ||
CR760236.2 | - | 7 - 8 | 1087 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |