Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203953.1 | NP_989284.1 | 8 - 9 | 1154 | 76 | |
GenBank mRNA | BC064221.1 | - | 8 - 9 | 1154 | 76 | |
CR761814.2 | - | 8 - 9 | 1136 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |