Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203957.1 | NP_989288.1 | 3 - 4 | 573 | 67 | |
GenBank mRNA | BC064179.1 | - | 3 - 4 | 573 | 67 | |
CR761345.2 | - | 3 - 4 | 465 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |