Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203969.1 | NP_989300.1 | 1 - 2 | 329 | 87 | |
GenBank mRNA | BC063917.1 | - | 1 - 2 | 329 | 87 | |
BC076710.1 | - | 1 - 2 | 363 | 87 | ||
CT030284.1 | - | 1 - 2 | 305 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |