Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc16a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203979.1 | NP_989310.1 | 574 | 75 | ||
GenBank mRNA | BC064255.1 | - | 574 | 75 | ||
CR855712.2 | - | 638 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |