Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203980.1 | NP_989311.1 | 4 - 5 | 696 | 59 | |
GenBank mRNA | BC063926.1 | - | 4 - 5 | 696 | 59 | |
CR926297.2 | - | 4 - 5 | 748 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |