Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203993.1 | NP_989324.1 | 4 - 5 | 595 | 100 | |
XM_018093825.1 | XP_017949314.1 | 4 - 5 | 639 | 100 | ||
XM_018093826.1 | XP_017949315.1 | 4 - 5 | 613 | 100 | ||
GenBank mRNA | BC064180.1 | - | 4 - 5 | 595 | 100 | |
CR761762.2 | - | 4 - 5 | 586 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |