Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203996.1 | NP_989327.1 | 2 - 3 | 162 | 75 | |
XM_012958472.2 | XP_012813926.1 | 2 - 3 | 255 | 75 | ||
GenBank mRNA | BC064170.1 | - | 2 - 3 | 162 | 75 | |
CR760855.2 | - | 2 - 3 | 106 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |