Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204000.1 | NP_989331.1 | 4 - 5 | 579 | 98 | |
XM_012956676.2 | XP_012812130.1 | 4 - 5 | 634 | 98 | ||
XM_012956677.2 | XP_012812131.1 | 4 - 5 | 651 | 98 | ||
XM_012956679.2 | XP_012812133.1 | 4 - 5 | 542 | 98 | ||
GenBank mRNA | BC064156.1 | - | 4 - 5 | 579 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |