Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204044.1 | NP_989375.1 | 1 - 2 | 44 | 81 | |
GenBank mRNA | BC064184.1 | - | 1 - 2 | 44 | 81 | |
CR762229.2 | - | 1 - 2 | 47 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |