Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204046.1 | NP_989377.1 | 10 - 11 | 1348 | 86 | |
XM_012971610.2 | XP_012827064.1 | 10 - 11 | 1403 | 86 | ||
GenBank mRNA | BC064723.1 | - | 10 - 11 | 1348 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |