Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hhex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204089.1 | NP_989420.1 | 1 - 2 | 408 | 93 | |
GenBank mRNA | AY160969.1 | - | 1 - 2 | 408 | 93 | |
BC171323.1 | - | 1 - 2 | 93 | |||
CR761571.2 | - | 1 - 2 | 408 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |