Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hhex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204089.1 | NP_989420.1 | 3 - 4 | 638 | 107 | |
GenBank mRNA | AY160969.1 | - | 3 - 4 | 638 | 107 | |
BC171323.1 | - | 3 - 4 | 635 | 107 | ||
CR761571.2 | - | 3 - 4 | 638 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |