Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tuba4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213682.1 | NP_998847.1 | 2 - 3 | 302 | 92 | |
GenBank mRNA | BC067974.1 | - | 2 - 3 | 302 | 92 | |
CR760596.2 | - | 1 - 2 | 92 | |||
CR926235.1 | - | 1 - 2 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |