Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213698.1 | NP_998863.1 | 7 - 8 | 1054 | 86 | |
XM_012962860.2 | XP_012818314.1 | 7 - 8 | 1029 | 86 | ||
GenBank mRNA | BC067913.1 | - | 7 - 8 | 1054 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |