Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pam16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004771.2 | NP_001004771.1 | 2 - 3 | 187 | 78 | |
GenBank mRNA | BC084449.1 | - | 2 - 3 | 187 | 78 | |
CR761515.2 | - | 2 - 3 | 137 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |