Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005116.2 | NP_001005116.1 | 364 | 65 | ||
GenBank mRNA | BC077055.1 | - | 366 | 65 | ||
CR760103.2 | - | 355 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |