Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005142.1 | NP_001005142.1 | 3 - 4 | 390 | 88 | |
GenBank mRNA | BC077690.1 | - | 3 - 4 | 390 | 88 | |
CT025161.2 | - | 3 - 4 | 288 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |