Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005455.1 | NP_001005455.1 | 4 - 5 | 461 | 78 | |
GenBank mRNA | BC074571.1 | - | 4 - 5 | 461 | 78 | |
CR855832.2 | - | 4 - 5 | 407 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |