Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005643.1 | NP_001005643.1 | 8 - 9 | 1366 | 70 | |
XM_012954970.2 | XP_012810424.1 | 8 - 9 | 1126 | 70 | ||
XM_012954971.2 | XP_012810425.1 | 8 - 9 | 1122 | 70 | ||
XM_012954972.2 | XP_012810426.1 | 8 - 9 | 1095 | 70 | ||
GenBank mRNA | BC074643.1 | - | 8 - 9 | 1366 | 70 | |
CR760378.2 | - | 7 - 8 | 1031 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |