Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005672.1 | NP_001005672.1 | 7 - 8 | 845 | 68 | |
GenBank mRNA | BC074710.1 | - | 7 - 8 | 845 | 68 | |
CR848543.2 | - | 7 - 8 | 806 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |