Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005691.1 | NP_001005691.1 | 1 - 2 | 1124 | 90 | |
GenBank mRNA | BC075118.1 | - | 1 - 2 | 1124 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |