Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: napsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005701.1 | NP_001005701.1 | 369 | 75 | ||
GenBank mRNA | BC075272.1 | - | 369 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |