Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cadm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005713.1 | NP_001005713.1 | 1401 | 56 | ||
GenBank mRNA | BC075300.1 | - | 1401 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis |