Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005818.1 | NP_001005818.1 | 1 - 2 | 268 | 84 | |
XM_018092061.1 | XP_017947550.1 | 1 - 2 | 152 | 84 | ||
XM_018092063.1 | XP_017947552.1 | 1 - 2 | 101 | 84 | ||
GenBank mRNA | BC075367.1 | - | 1 - 2 | 268 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |