Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: krt70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006686.1 | NP_001006687.1 | 3 - 4 | 642 | 89 | |
GenBank mRNA | BC074628.1 | - | 3 - 4 | 642 | 89 | |
CR760223.2 | - | 3 - 4 | 629 | 89 | ||
CT485755.1 | - | 3 - 4 | 632 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |