Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006695.1 | NP_001006696.1 | 4 - 5 | 505 | 104 | |
GenBank mRNA | BC075400.1 | - | 4 - 5 | 505 | 104 | |
CR761026.2 | - | 4 - 5 | 479 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |