Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006717.2 | NP_001006718.1 | 6 - 7 | 554 | 102 | |
XM_012969759.2 | XP_012825213.1 | 5 - 6 | 500 | 102 | ||
GenBank mRNA | BC075449.1 | - | 6 - 7 | 556 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |