Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006747.1 | NP_001006748.1 | 3 - 4 | 378 | 96 | |
GenBank mRNA | BC075516.1 | - | 3 - 4 | 378 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |