Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006751.1 | NP_001006752.1 | 2 - 3 | 532 | 70 | |
GenBank mRNA | BC075524.1 | - | 2 - 3 | 532 | 70 | |
CT030462.1 | - | 2 - 3 | 150 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |