Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006754.1 | NP_001006755.1 | 4 - 5 | 1418 | 70 | |
GenBank mRNA | BC075531.1 | - | 4 - 5 | 1418 | 70 |
Gene Symbol: LOC100489903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018094467.1 | XP_017949956.1 | 4 - 5 | 1446 | 70 | |
XM_018094468.1 | XP_017949957.1 | 4 - 5 | 1295 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |