Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spa17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006757.1 | NP_001006758.1 | 2 - 3 | 245 | 83 | |
XM_012966399.2 | XP_012821853.1 | 2 - 3 | 516 | 83 | ||
XM_012966400.2 | XP_012821854.1 | 2 - 3 | 631 | 83 | ||
XM_012966402.2 | XP_012821856.1 | 2 - 3 | 520 | 83 | ||
GenBank mRNA | BC075539.1 | - | 2 - 3 | 245 | 83 | |
CT030312.1 | - | 295 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |