Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsnax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006777.1 | NP_001006778.1 | 3 - 4 | 392 | 93 | |
GenBank mRNA | BC075580.1 | - | 3 - 4 | 392 | 93 | |
CR942685.1 | - | 3 - 4 | 254 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |