Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006778.1 | NP_001006779.1 | 2 - 3 | 320 | 64 | |
GenBank mRNA | BC075582.1 | - | 2 - 3 | 320 | 64 | |
CR761718.2 | - | 2 - 3 | 385 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |