Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006811.1 | NP_001006812.1 | 2 - 3 | 427 | 133 | |
GenBank mRNA | BC076692.1 | - | 2 - 3 | 427 | 133 | |
CR762126.2 | - | 2 - 3 | 381 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |